6-Bromo-2-trifluoromethyl-1H-indole
Dodavatel: Apollo Scientific
|
Varování
|
PC421113-1G
PC421113-250MG
PC421113-5G
APOSPC421113-1GEA
11740
CZK
APOSPC421113-1G
APOSPC421113-250MG
APOSPC421113-5G
6-Bromo-2-trifluoromethyl-1H-indole
6-Bromo-2-trifluoromethyl-1H-indole
Vzorec:
C₉H₅BrF₃N MW: 264,042 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD30721252 CAS číslo: 1782390-81-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...