Tisk…

5-Bromo-2-(trifluoromethyl)isonicotinonitrile

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Bromo-2-(trifluoromethyl)pyridine-4-carbonitrile

PC421042-100MG PC421042-250MG
APOSPC421042-100MGEA 2530 CZK
APOSPC421042-100MG APOSPC421042-250MG
5-Bromo-2-(trifluoromethyl)isonicotinonitrile
5-Bromo-2-(trifluoromethyl)isonicotinonitrile
Vzorec: C₇H₂BrF₃N₂
MW: 251,003 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18261445
CAS číslo: 1070892-04-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR