Tisk…

2-Bromo-5-(difluoromethoxy)pyridine

Dodavatel: Apollo Scientific
Varování

PC421030-1G PC421030-500MG PC421030-250MG
APOSPC421030-1GEA 2240 CZK
APOSPC421030-1G APOSPC421030-500MG APOSPC421030-250MG
2-Bromo-5-(difluoromethoxy)pyridine
2-Bromo-5-(difluoromethoxy)pyridine
Vzorec: C₆H₄BrF₂NO
MW: 224,003 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185842
CAS číslo: 845827-14-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR