Tisk…

2-Chloro-6-(difluoromethoxy)pyridine

Dodavatel: Apollo Scientific
Varování

PC421028-1G PC421028-500MG
APOSPC421028-1GEA 8570 CZK
APOSPC421028-1G APOSPC421028-500MG
2-Chloro-6-(difluoromethoxy)pyridine
2-Chloro-6-(difluoromethoxy)pyridine
Vzorec: C₆H₄ClF₂NO
MW: 179.552 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185850
CAS číslo: 1214377-45-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR