Tisk…

2',4'-Dichloro-2-(4-fluorophenyl)acetophenone

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 1-(2,4-Dichlorophenyl)-2-(4-fluorophenyl)ethanone

PC421010-1G PC421010-250MG PC421010-5G
APOSPC421010-1GEA 3940 CZK
APOSPC421010-1G APOSPC421010-250MG APOSPC421010-5G
2',4'-Dichloro-2-(4-fluorophenyl)acetophenone
2',4'-Dichloro-2-(4-fluorophenyl)acetophenone
Vzorec: C₁₄H₉Cl₂FO
MW: 283,125 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16350604
CAS číslo: 611220-24-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR