4'-Chloro-2-(3,4-difluorophenyl)acetophenone
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC421001-1G
PC421001-250MG
PC421001-5G
APOSPC421001-1GEA
3940
CZK
APOSPC421001-1G
APOSPC421001-250MG
APOSPC421001-5G
4'-Chloro-2-(3,4-difluorophenyl)acetophenone
4'-Chloro-2-(3,4-difluorophenyl)acetophenone
Vzorec:
C₁₄H₉ClF₂O Storage Temperature: Ambient |
MDL Number:
MFCD23417249 CAS číslo: 1373156-53-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...