[2-Chloro-6-(trifluoromethyl)pyridin-4-yl]boronic acid 95%
Dodavatel: Apollo Scientific
Synonyms:
(2-Chloro-6-(trifluoromethyl)pyridin-4-yl)boronic acid, 2-Chloro-6-(trifluoromethyl)pyridine-4-boronic acid
PC412554-1G
PC412554-250MG
APOSPC412554-1GEA
5100
CZK
APOSPC412554-1G
APOSPC412554-250MG
[2-Chloro-6-(trifluoromethyl)pyridin-4-yl]boronic acid 95%
[2-Chloro-6-(trifluoromethyl)pyridin-4-yl]boronic acid
Vzorec:
C₆H₄BClF₃NO₂ MW: 225.361 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13188845 CAS číslo: 1446486-10-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...