Tisk…

3-Methyl-4-trifluoromethoxyphenylboronic acid

Dodavatel: Apollo Scientific

Synonyms: (3-Methyl-4-(trifluoromethoxy)phenyl)boronic acid

PC412525-1G PC412525-250MG
APOSPC412525-1GEA 10550 CZK
APOSPC412525-1G APOSPC412525-250MG
3-Methyl-4-trifluoromethoxyphenylboronic acid
3-Methyl-4-trifluoromethoxyphenylboronic acid
Vzorec: C₈H₈BF₃O₃
MW: 219,954 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11504858
CAS číslo: 871362-79-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR