Tisk…

5-Hydroxy-2-(trifluoromethoxy)phenylboronic acid

Dodavatel: Apollo Scientific

PC412498-1G PC412498-5G
APOSPC412498-1GEA 7810 CZK
APOSPC412498-1G APOSPC412498-5G
5-Hydroxy-2-(trifluoromethoxy)phenylboronic acid
5-Hydroxy-2-(trifluoromethoxy)phenylboronic acid
Vzorec: C₇H₆BF₃O₄
MW: 221,926 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12913937
CAS číslo: 1217500-69-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR