Tisk…

[2-Fluoro-4-(hydroxymethyl)phenyl]boronic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: (2-Fluoro-4-(hydroxymethyl)phenyl)boronic acid, 2-Fluoro-4-(hydroxymethyl)benzeneboronic acid

PC412422-1G PC412422-5G
APOSPC412422-1GEA 3540 CZK
APOSPC412422-1G APOSPC412422-5G
[2-Fluoro-4-(hydroxymethyl)phenyl]boronic acid
[2-Fluoro-4-(hydroxymethyl)phenyl]boronic acid
Vzorec: C₇H₈BFO₃
MW: 169,946 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD21603758
CAS číslo: 1331945-14-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR