Tisk…

2,3-Difluoropyridine-4-boronic acid 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: (2,3-Difluoropyridin-4-yl)boronic acid

PC412378-5G
APOSPC412378-5GEA 4370 CZK
APOSPC412378-5G
2,3-Difluoropyridine-4-boronic acid 97%
2,3-Difluoropyridine-4-boronic acid
Vzorec: C₅H₄BF₂NO₂
MW: 158,899 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12022339
CAS číslo: 1263374-42-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR