Tisk…

2,4-Difluoro-5-methoxyphenylboronic acid 98%

Dodavatel: Apollo Scientific
Varování

PC412365-1G PC412365-250MG
APOSPC412365-1GEA 11580 CZK
APOSPC412365-1G APOSPC412365-250MG
2,4-Difluoro-5-methoxyphenylboronic acid 98%
2,4-Difluoro-5-methoxyphenylboronic acid
Vzorec: C₇H₇BF₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD21609510
CAS číslo: 1395417-65-8

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR