Tisk…

2,4-Dichloro-5-fluorophenylboronic acid 98%

Dodavatel: Apollo Scientific
Varování

PC412347-1G PC412347-250MG
APOSPC412347-1GEA 24170 CZK
APOSPC412347-1G APOSPC412347-250MG
2,4-Dichloro-5-fluorophenylboronic acid 98%
2,4-Dichloro-5-fluorophenylboronic acid
Vzorec: C₆H₄BCl₂FO₂
Storage Temperature: Refrigerator
MDL Number: MFCD19981520
CAS číslo: 1417914-09-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR