3-Chloro-4-(trifluoromethoxy)phenylboronic acid
Dodavatel: Apollo Scientific
|
Varování
|
PC412324-1G
PC412324-250MG
APOSPC412324-1GEA
20440
CZK
APOSPC412324-1G
APOSPC412324-250MG
3-Chloro-4-(trifluoromethoxy)phenylboronic acid
3-Chloro-4-(trifluoromethoxy)phenylboronic acid
Vzorec:
C₇H₅BClF₃O₃ MW: 240,372 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD10697423 CAS číslo: 870822-79-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...