2-Fluoro-5-isopropylphenylboronic acid
Dodavatel: Apollo Scientific
Synonyms:
(2-Fluoro-5-isopropylphenyl)boronic acid
PC412198-1G
PC412198-250MG
APOSPC412198-1GEA
12240
CZK
APOSPC412198-1G
APOSPC412198-250MG
2-Fluoro-5-isopropylphenylboronic acid
2-Fluoro-5-isopropylphenylboronic acid
Vzorec:
C₉H₁₂BFO₂ MW: 182 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD17015760 CAS číslo: 1256354-92-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...