Tisk…

3,5-Difluoro-4-methoxybenzeneboronic acid 96%

Dodavatel: Apollo Scientific
Varování

PC412069-1G PC412069-500MG PC412069-5G
APOSPC412069-1GEA 4370 CZK
APOSPC412069-1G APOSPC412069-500MG APOSPC412069-5G
3,5-Difluoro-4-methoxybenzeneboronic acid 96%
3,5-Difluoro-4-methoxybenzeneboronic acid
Vzorec: C₇H₇BF₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD08706257
CAS číslo: 208641-98-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR