4-Chloro-1-(2-fluoroethyl)-1H-pyrazole 95+%
Dodavatel: Apollo Scientific
|
Varování
|
PC411327-1G
PC411327-5G
APOSPC411327-1GEA
2480
CZK
APOSPC411327-1G
APOSPC411327-5G
4-Chloro-1-(2-fluoroethyl)-1H-pyrazole 95+%
4-Chloro-1-(2-fluoroethyl)-1H-pyrazole
Vzorec:
C₅H₆ClFN₂ Storage Temperature: Ambient |
MDL Number:
MFCD25371345 CAS číslo: 1429419-75-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...