Tisk…

1-[3-(Difluoromethoxy)phenyl]methanamine hydrochloride

Dodavatel: Apollo Scientific
Varování

PC411323-1G PC411323-5G
APOSPC411323-1GEA 1330 CZK
APOSPC411323-1G APOSPC411323-5G
1-[3-(Difluoromethoxy)phenyl]methanamine hydrochloride
1-[3-(Difluoromethoxy)phenyl]methanamine hydrochloride
Vzorec: C₈H₉F₂NO·ClH
MW: 209,621 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15209619
CAS číslo: 1106917-71-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR