2-(Difluoromethoxy)-5-fluoroaniline hydrochloride
Dodavatel: Apollo Scientific
|
Varování
|
PC411308-1G
PC411308-5G
APOSPC411308-1GEA
1040
CZK
APOSPC411308-1G
APOSPC411308-5G
2-(Difluoromethoxy)-5-fluoroaniline hydrochloride
2-(Difluoromethoxy)-5-fluoroaniline hydrochloride
Vzorec:
C₇H₆F₃NO·ClH Storage Temperature: Refrigerator |
MDL Number:
MFCD25368949 CAS číslo: 1431966-19-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...