Tisk…

1-(2-Fluoroethyl)-4-iodo-3-methyl-1H-pyrazole

Dodavatel: Apollo Scientific
Varování

PC411305-1G PC411305-5G
APOSPC411305-1GEA 870 CZK
APOSPC411305-1G APOSPC411305-5G
1-(2-Fluoroethyl)-4-iodo-3-methyl-1H-pyrazole
1-(2-Fluoroethyl)-4-iodo-3-methyl-1H-pyrazole
Vzorec: C₆H₈FIN₂
MW: 254,044 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25949444
CAS číslo: 1443278-85-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR