Tisk…

2-Fluoro-7-azaspiro[3.5]nonane hydrochloride

Dodavatel: Apollo Scientific

PC411285-1G PC411285-250MG PC411285-500MG
APOSPC411285-1GEA 49260 CZK
APOSPC411285-1G APOSPC411285-250MG APOSPC411285-500MG
2-Fluoro-7-azaspiro[3.5]nonane hydrochloride
2-Fluoro-7-azaspiro[3.5]nonane hydrochloride
Vzorec: C₈H₁₄FN·ClH
MW: 179.663 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22683335
CAS číslo: 1263178-15-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR