Tisk…
2-Chloro-2,2-difluoroethanesulphonyl fluoride 97%
Dodavatel: Apollo Scientific
PC4110-10G PC4110-50G
APOSPC4110-10GEA 10350 CZK
APOSPC4110-10G APOSPC4110-50G
2-Chloro-2,2-difluoroethanesulphonyl fluoride 97%
2-Chloro-2,2-difluoroethanesulphonyl fluoride
Vzorec: C₂H₂ClF₃O₂S
MW: 182,549 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03412192
CAS číslo: 460-94-6