Tisk…

N-t-BOC-cis-4-Fluoro-L-prolinonitrile

Dodavatel: Apollo Scientific

Synonyms: (2S,4S)-tert-Butyl 2-cyano-4-fluoropyrrolidine-1-carboxylate

PC409010-1G PC409010-250MG
APOSPC409010-1GEA 3190 CZK
APOSPC409010-1G APOSPC409010-250MG
N-t-BOC-cis-4-Fluoro-L-prolinonitrile
N-t-BOC-cis-4-Fluoro-L-prolinonitrile
Vzorec: C₁₀H₁₅FN₂O₂
MW: 214,237 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09263600
CAS číslo: 426844-76-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR