Tisk…

2-Fluoropyrimidin-4-amine

Dodavatel: Apollo Scientific

PC408561-1G PC408561-250MG
APOSPC408561-1GEA 17530 CZK
APOSPC408561-1G APOSPC408561-250MG
2-Fluoropyrimidin-4-amine
2-Fluoropyrimidin-4-amine
Vzorec: C₄H₄FN₃
MW: 113,093 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00769380
CAS číslo: 96548-91-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR