Tisk…

Methyl difluoro(trimethylsilyl)acetate 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC408185-1G PC408185-25G PC408185-5G
APOSPC408185-1GEA 2280 CZK
APOSPC408185-1G APOSPC408185-25G APOSPC408185-5G
Methyl difluoro(trimethylsilyl)acetate 97%
Methyl difluoro(trimethylsilyl)acetate
Vzorec: C₆H₁₂F₂O₂Si
MW: 182,241 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04973091
CAS číslo: 730971-66-1
UN: 1993
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR