Tisk…

Ethyl 3-chloro-4,4,4-trifluorobutyrate

Dodavatel: Apollo Scientific

PC408107-25G PC408107-5G
APOSPC408107-25GEA 33060 CZK
APOSPC408107-25G APOSPC408107-5G
Ethyl 3-chloro-4,4,4-trifluorobutyrate
Ethyl 3-chloro-4,4,4-trifluorobutyrate
Vzorec: C₆H₈ClF₃O₂
MW: 204,575 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18641867
CAS číslo: 1309602-63-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR