Ethyl 2,2,3,3-tetrafluoropropionate
Dodavatel: Apollo Scientific
|
Varování
|
PC408104-25G
PC408104-5G
PC408104-100G
APOSPC408104-25GEA
8570
CZK
APOSPC408104-25G
APOSPC408104-5G
APOSPC408104-100G
Ethyl 2,2,3,3-tetrafluoropropionate
Ethyl 2,2,3,3-tetrafluoropropionate
Vzorec:
C₅H₆F₄O₂ MW: 174,094 g/mol Bod varu: 107…109 °C Hustota: 1,28 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD18641870 CAS číslo: 337-82-6 UN: 3272 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...