2,2,3,4,4,4-Hexafluorobutyric acid 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC408093-1G
PC408093-5G
APOSPC408093-1GEA
4900
CZK
APOSPC408093-1G
APOSPC408093-5G
2,2,3,4,4,4-Hexafluorobutyric acid 97%
2,2,3,4,4,4-Hexafluorobutyric acid
Vzorec:
C₄H₂F₆O₂ Bod varu: 140 °C (740 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD16621311 CAS číslo: 379-90-8 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...