8-Fluorosulphonylperfluoro(2,5-dimethyl-3,6-dioxaoctanoyl) fluoride 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC408082-100G
PC408082-50G
APOSPC408082-100GEA
23150
CZK
APOSPC408082-100G
APOSPC408082-50G
8-Fluorosulphonylperfluoro(2,5-dimethyl-3,6-dioxaoctanoyl) fluoride 95%
8-Fluorosulphonylperfluoro(2,5-dimethyl-3,6-dioxaoctanoyl) fluoride
Vzorec:
C₈F₁₆O₅S MW: 512,122 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16621332 CAS číslo: 4089-58-1 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...