Tisk…

tert-Butyl 2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenylcarbamate

Dodavatel: Apollo Scientific

Synonyms: 4-(tert-Butoxycarbonylamino)-3-fluorophenylboronic acid, pinacol ester

PC408014-1G PC408014-250MG PC408014-5G
APOSPC408014-1GEA 8100 CZK
APOSPC408014-1G APOSPC408014-250MG APOSPC408014-5G
tert-Butyl 2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenylcarbamate
tert-Butyl 2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenylcarbamate
Vzorec: C₁₇H₂₅BFNO₄
MW: 337,194 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09033883
CAS číslo: 262444-42-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR