Tisk…

4-Bromo-5-fluoro-1,2-dihydroisoquinolin-1-one

Dodavatel: Apollo Scientific
Nebezpečí

PC407085-100MG PC407085-1G PC407085-500MG
APOSPC407085-100MGEA 9960 CZK
APOSPC407085-100MG APOSPC407085-1G APOSPC407085-500MG
4-Bromo-5-fluoro-1,2-dihydroisoquinolin-1-one
4-Bromo-5-fluoro-1,2-dihydroisoquinolin-1-one
Vzorec: C₉H₅BrFNO
Storage Temperature: Ambient
MDL Number: MFCD15832833
CAS číslo: 1207448-50-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR