Tisk…

8-Fluoro-4-hydrazinoquinoline hydrochloride

Dodavatel: Apollo Scientific

PC407066-1G PC407066-5G
APOSPC407066-1GEA 8830 CZK
APOSPC407066-1G APOSPC407066-5G
8-Fluoro-4-hydrazinoquinoline hydrochloride
8-Fluoro-4-hydrazinoquinoline hydrochloride
Vzorec: C₉H₈FN₃·ClH
MW: 213,639 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02684142
CAS číslo: 1171070-31-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR