Tisk…

Methyl (1S,2R,3R,8S)-4-fluorocubane-1-carboxylate 95%

Dodavatel: Apollo Scientific

PC405712-1G PC405712-250MG
APOSPC405712-1GEA 128610 CZK
APOSPC405712-1G APOSPC405712-250MG
Methyl (1S,2R,3R,8S)-4-fluorocubane-1-carboxylate 95%
Methyl (1S,2R,3R,8S)-4-fluorocubane-1-carboxylate
Vzorec: C₁₀H₉FO₂
MW: 180.176 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30723461
CAS číslo: 141046-54-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR