Tisk…

Methyl (1S,2R,3R,8S)-4-(fluoromethyl)cubane-1-carboxylate 95%

Dodavatel: Apollo Scientific

PC405711-1G PC405711-250MG
APOSPC405711-1GEA 118360 CZK
APOSPC405711-1G APOSPC405711-250MG
Methyl (1S,2R,3R,8S)-4-(fluoromethyl)cubane-1-carboxylate 95%
Methyl (1S,2R,3R,8S)-4-(fluoromethyl)cubane-1-carboxylate
Vzorec: C₁₁H₁₁FO₂
MW: 194,202 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30723460
CAS číslo: 1350822-06-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR