Tisk…

(1S,2R,3R,8S)-4-(Fluoromethyl)cubane-1-carboxylic acid 95%

Dodavatel: Apollo Scientific

PC405709-1G PC405709-250MG
APOSPC405709-1GEA 128610 CZK
APOSPC405709-1G APOSPC405709-250MG
(1S,2R,3R,8S)-4-(Fluoromethyl)cubane-1-carboxylic acid 95%
(1S,2R,3R,8S)-4-(Fluoromethyl)cubane-1-carboxylic acid
Vzorec: C₁₀H₉FO₂
Storage Temperature: Ambient
MDL Number: MFCD24502265
CAS číslo: 1350821-83-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR