Tisk…

((1S,2R,3R,8S)-4-Fluorocuban-1-yl)methanol 95%

Dodavatel: Apollo Scientific

PC405708-1G PC405708-250MG
APOSPC405708-1GEA 128610 CZK
APOSPC405708-1G APOSPC405708-250MG
((1S,2R,3R,8S)-4-Fluorocuban-1-yl)methanol 95%
((1S,2R,3R,8S)-4-Fluorocuban-1-yl)methanol
Vzorec: C₉H₉FO
MW: 152,166 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28955862
CAS číslo: 167496-73-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR