Tisk…

1-(3-Fluorobicyclo[1.1.1]pentan-1-yl)ethan-1-one

Dodavatel: Apollo Scientific
Varování

PC405700-1G PC405700-250MG
APOSPC405700-1GEA 118030 CZK
APOSPC405700-1G APOSPC405700-250MG
1-(3-Fluorobicyclo[1.1.1]pentan-1-yl)ethan-1-one
1-(3-Fluorobicyclo[1.1.1]pentan-1-yl)ethan-1-one
Vzorec: C₇H₉FO
Storage Temperature: Refrigerator
MDL Number: MFCD24709482
CAS číslo: 262852-00-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR