Ethyl 3-fluorobicyclo[1.1.1]pentane-1-carboxylate
Dodavatel: Apollo Scientific
PC405697-1G
PC405697-250MG
APOSPC405697-1GEA
130260
CZK
APOSPC405697-1G
APOSPC405697-250MG
Ethyl 3-fluorobicyclo[1.1.1]pentane-1-carboxylate
Ethyl 3-fluorobicyclo[1.1.1]pentane-1-carboxylate
Vzorec:
C₈H₁₁FO₂ Storage Temperature: Ambient |
MDL Number:
MFCD29037344 CAS číslo: 1935130-91-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...