4-(3-Fluorobicyclo[1.1.1]pentan-1-yl)aniline 95%
Dodavatel: Apollo Scientific
PC405693-1G
PC405693-250MG
APOSPC405693-1GEA
88940
CZK
APOSPC405693-1G
APOSPC405693-250MG
4-(3-Fluorobicyclo[1.1.1]pentan-1-yl)aniline 95%
4-(3-Fluorobicyclo[1.1.1]pentan-1-yl)aniline
Vzorec:
C₁₁H₁₂FN Storage Temperature: Ambient |
MDL Number:
MFCD29037340 CAS číslo: 1936606-29-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...