Tisk…

3-Fluorobicyclo[1.1.1]pentane-1-carboxylic acid 97%

Dodavatel: Apollo Scientific

PC405690-1G PC405690-250MG PC405690-5G
APOSPC405690-1GEA 6290 CZK
APOSPC405690-1G APOSPC405690-250MG APOSPC405690-5G
3-Fluorobicyclo[1.1.1]pentane-1-carboxylic acid 97%
3-Fluorobicyclo[1.1.1]pentane-1-carboxylic acid
Vzorec: C₆H₇FO₂
Storage Temperature: Ambient
MDL Number: MFCD19228680
CAS číslo: 146038-53-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR