Tisk…

1-(Pentafluorosulphanyl)-3-bromo-5-nitrobenzene

Dodavatel: Apollo Scientific
Nebezpečí

PC405636-1G PC405636-250MG
APOSPC405636-1GEA 17990 CZK
APOSPC405636-1G APOSPC405636-250MG
1-(Pentafluorosulphanyl)-3-bromo-5-nitrobenzene
1-(Pentafluorosulphanyl)-3-bromo-5-nitrobenzene
Vzorec: C₆H₃BrF₅NO₂S
Storage Temperature: Ambient
MDL Number: MFCD27664831
CAS číslo: 871507-63-0
UN: 2811
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR