2-Nitro-5-(pentafluorosulphanyl)phenol
Dodavatel: Apollo Scientific
|
Varování
|
PC405530-1G
PC405530-250MG
APOSPC405530-1GEA
9920
CZK
APOSPC405530-1G
APOSPC405530-250MG
2-Nitro-5-(pentafluorosulphanyl)phenol
2-Nitro-5-(pentafluorosulphanyl)phenol
Vzorec:
C₆H₄F₅NO₃S MW: 265,158 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22370112 CAS číslo: 1329120-21-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...