Tisk…

3-Bromo-8-fluoro-4-hydroxyquinoline

Dodavatel: Apollo Scientific

Synonyms: 3-Bromo-8-fluoroquinolin-4-ol

PC404591-1G PC404591-250MG
APOSPC404591-1GEA 10250 CZK
APOSPC404591-1G APOSPC404591-250MG
3-Bromo-8-fluoro-4-hydroxyquinoline
3-Bromo-8-fluoro-4-hydroxyquinoline
Vzorec: C₉H₅BrFNO
MW: 242,045 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13192868
CAS číslo: 1065087-83-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR