4-Bromo-6,8-difluoro-2-methylquinoline
Dodavatel: Apollo Scientific
PC404590-1G
PC404590-5G
APOSPC404590-1GEA
0
CZK
APOSPC404590-1G
APOSPC404590-5G
4-Bromo-6,8-difluoro-2-methylquinoline
4-Bromo-6,8-difluoro-2-methylquinoline
Vzorec:
C₁₀H₆BrF₂N Storage Temperature: Ambient |
MDL Number:
MFCD12675031 CAS číslo: 1189106-86-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...