4-Amino-6,8-difluoro-2-methylquinoline
Dodavatel: Apollo Scientific
PC404575-1G
PC404575-5G
APOSPC404575-1GEA
8100
CZK
APOSPC404575-1G
APOSPC404575-5G
4-Amino-6,8-difluoro-2-methylquinoline
4-Amino-6,8-difluoro-2-methylquinoline
Vzorec:
C₁₀H₈F₂N₂ Storage Temperature: Ambient |
MDL Number:
MFCD11053597 CAS číslo: 288151-32-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...