Tisk…
2,8-Bis(trifluoromethyl)-4-quinolyl(1-oxypyrid-2-yl) methane
Dodavatel: Apollo Scientific
PC404545-1G PC404545-5G
APOSPC404545-1GEA 8570 CZK
APOSPC404545-1G APOSPC404545-5G
2,8-Bis(trifluoromethyl)-4-quinolyl(1-oxypyrid-2-yl) methane
2,8-Bis(trifluoromethyl)-4-quinolyl(1-oxypyrid-2-yl) methane
Vzorec: C₁₇H₁₀F₆N₂O
MW: 372.265 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10698487
CAS číslo: 83012-10-6