2-Amino-6-fluoro-3-methylquinoline
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
6-Fluoro-3-methylquinolin-2-amine
PC404500-1G
PC404500-5G
APOSPC404500-1GEA
9530
CZK
APOSPC404500-1G
APOSPC404500-5G
2-Amino-6-fluoro-3-methylquinoline
2-Amino-6-fluoro-3-methylquinoline
Vzorec:
C₁₀H₉FN₂ MW: 176,19 g/mol Bod tání: 176…178 °C Storage Temperature: Ambient |
MDL Number:
MFCD00272512 CAS číslo: 203506-28-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...