Tisk…

1-(2-Fluoroethyl)-1H-pyrazole

Dodavatel: Apollo Scientific
Varování

PC404014-1G PC404014-25G PC404014-5G
APOSPC404014-1GEA 622 CZK
APOSPC404014-1G APOSPC404014-25G APOSPC404014-5G
1-(2-Fluoroethyl)-1H-pyrazole
1-(2-Fluoroethyl)-1H-pyrazole
Vzorec: C₅H₇FN₂
MW: 114.121 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23972194
CAS číslo: 139578-67-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR