2-Fluoro-5-methoxypyridine 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC404001-10G
PC404001-1G
PC404001-5G
APOSPC404001-10GEA
10650
CZK
APOSPC404001-10G
APOSPC404001-1G
APOSPC404001-5G
2-Fluoro-5-methoxypyridine 98%
2-Fluoro-5-methoxypyridine
Vzorec:
C₆H₆FNO MW: 127.116 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13185604 CAS číslo: 136888-79-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...