Tisk…

(6-Fluoropyridin-3-yl)methylamine

Dodavatel: Apollo Scientific

Synonyms: (6-Fluoropyridin-3-yl)methanamine

PC403115-1G PC403115-250MG PC403115-5G
APOSPC403115-1GEA 4270 CZK
APOSPC403115-1G APOSPC403115-250MG APOSPC403115-5G
(6-Fluoropyridin-3-yl)methylamine
(6-Fluoropyridin-3-yl)methylamine
Vzorec: C₆H₇FN₂
MW: 126,132 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11100747
CAS číslo: 205744-17-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR